Anti-TP53BP1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-864
10074-450EA 582.9 USD
10074-450
Anti-TP53BP1 Rabbit Polyclonal Antibody
Antibodies
P53-binding protein 1 (53BP1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, it is phosphorylated and relocalizes to the presumptive sites of damage, specifically, double-strand breaks. This also suggests a role in DNA repair, maintaining genomic stability. Tumor protein p53 binding protein 1 binds to the central domain of p53 required for site-specific DNA binding thus inhibiting the ability of p53 to bind DNA. TP53BP1 also functions as a checkpoint protein with properties of a DNA double-strand break sensor. It is a putative substrate of ATM kinase and is phosphorylated in response to DNA damage and localizes to sites of double-strand breaks.

TP53BP1 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

Affinity Chromatography

PBS, pH 7.2, no preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TP53BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR