ATM-3E8 recognizes full-length ATM, a 370 kDa nuclear phosphoprotein, which is involved in the autosomal recessive disease ataxia telangiectasia (AT). ATM belongs to a novel family of proteins associated with cell cycle regulation and response to DNA repair. The C-terminal region has extensive homology to the catalytic domains of phosphatidylinositol 3-kinases (PI3 kinases). Its usefulness to monitor altered ATM expression in cancer is under active investigation. The ATM gene product is a 370 kD nuclear phosphoprotein with C-terminal homology to phosphatidylinositol 3-kinase. ATM is involved in the induction of a DNA damage control pathway. Mutations in the ATM gene cause neuronal degeneration, gonadal abnormalities, and immune deficiencies as part of the hereditary disease ataxia telangiectasia.
ATM antibody can be used in ELISA starting at 1:200 - 1:1000, Western Blot starting at 1:100 - 1:300, immunocytochemistry starting at 1:75, and immunofluorescence starting at 1:25 - 1:50.
Protein G Column
PBS, pH 7.4. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: [3E8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human