Anti-STAT5B Rabbit Polyclonal Antibody

Supplier: Prosci

48-830
10078-370EA 561.89 USD InStock
10078-370
Anti-STAT5B Rabbit Polyclonal Antibody
Antibodies
STAT5B antibody can be used in ELISA, and immunohistochemistry starting at 15 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STAT5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR