Anti-PLK1 Rabbit Polyclonal Antibody

48-825
10078-364EA 674.53 USD
10078-364
Anti-PLK1 Rabbit Polyclonal Antibody
Antibodies
PLK, a Polo type protein kinase, has been shown to function in mitosis. The mRNA is absent or expressed at very low levels during the G1 phase, begins to reaccumulate during the S phase, and reaches maximal levels during the G2/M phase. PLK1 may participate in targeting MPF to the nucleus during prophase and in regulating mitotic spindle function. These functions, in addition to its expression in proliferating cells, suggest that PLK1 has a role in cancer progression.

Polo-Like Kinase antibody can be used in Western Blot, and immunohistochemistry starting at 20 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR