Anti-TIMP2 Mouse Monoclonal Antibody [clone: [F27P3A4]]

48-811
10074-414EA 698.16 USD
10074-414
Anti-TIMP2 Mouse Monoclonal Antibody [clone: [F27P3A4]]
Antibodies
Metalloproteinase Inhibitor 2 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone: [F27P3A4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Gibbon, Bovine, Chicken, Dog, Guinea pig, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR