Anti-WDHD1 Mouse Monoclonal Antibody [clone: [20G10]]

48-807
10074-406EA 698.16 USD
10074-406
Anti-WDHD1 Mouse Monoclonal Antibody [clone: [20G10]]
Antibodies
WDHD1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 20 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: WDHD1
Clonality: Monoclonal
Clone: [20G10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR