Anti-SP3 Rabbit Polyclonal Antibody

Supplier: Prosci

48-799
10076-052EA 582.9 USD
10076-052
Anti-SP3 Rabbit Polyclonal Antibody
Antibodies
SP3 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, and flow cytometry.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 0.2% gelatin.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: SP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR