Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MDM2 Mouse Monoclonal Antibody

Supplier: Prosci

48-781
10076-028EA 515.21 USD InStock
10076-028
Anti-MDM2 Mouse Monoclonal Antibody
Antibodies
This product requires antigen retrieval using heat treatment prior to staining of paraffin sections. EDTA pH8. is recommended for this purpose. IHC Control: Breast Carcinoma.

MDM2 antibody can be used in immunohistochemistry starting at 1:50, and flow cytometry starting at 1 µg/1E6 cells.

IgG Fractionation

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MDM2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Dog, Human, Monkey, Orangutan, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty