Anti-SLC45A3 Rabbit Polyclonal Antibody

48-774
10076-018EA 568.02 USD
10076-018
Anti-SLC45A3 Rabbit Polyclonal Antibody
Prostein antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC45A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Gibbon, Bovine, Chicken, Dog, Gorilla, Hamster, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR