Anti-MCL1 Mouse Monoclonal Antibody [clone: [8C6, 8C6D4B1]]

Supplier: Prosci

48-737
10074-358EA 582.9 USD
10074-358
Anti-MCL1 Mouse Monoclonal Antibody [clone: [8C6, 8C6D4B1]]
Antibodies
Myeloid cell leukimia-1 (Mcl-1) is a member of the Bcl-2 family of proteins that can act to promote cell survival. While the mechanism by which Mcl-1 inhibits apoptosis is not known, it is thought that it may heterodimerize and neutralize pro-apoptotic members of the Bcl-2 family such as Bim or Bak. Mcl-1 was originally identified in differentiating myeloid cells, but has since been shown to be expressed in multiple cell types. Mcl-1 is essential for embryogenesis and for the development and maintenance of B and T lymphocytes in animals. Mcl-1 exists as at least two distinct isoforms designated Mcl-1L and Mcl-1S. In marked contrast to the larger isoform of Mcl-1, overexpression of Mcl-1S promotes cell death.

MCL1 antibody can be used in ELISA, immunofluorescence, and immunohistochemistry starting at 0.31 ?g/mL.

Protein G Column

PBS, pH 7.4, 0.03% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MCL1
Clonality: Monoclonal
Clone: [8C6, 8C6D4B1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR