Anti-INHA Mouse Monoclonal Antibody

48-732
10074-348EA 684.46 USD
10074-348
Anti-INHA Mouse Monoclonal Antibody
Antibodies
Inhibin alpha antibody can be used in ELISA starting at 1:4000 - 1:20000, Western Blot starting at 1:250 - 1:1500, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.03% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: INHA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR