Anti-ACADM Rabbit Polyclonal Antibody

48-724
10078-352EA 684.46 USD
10078-352
Anti-ACADM Rabbit Polyclonal Antibody
Antibodies
ACADM encodes the medium-chain specific (C4 to C12 straight chain) Acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Clinical phenotypes are associated with ACADM hereditary deficiency.

ACADM antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry starting at 1:10 - 1:25.

Protein A Column

Tris buffered saline, 0.1% BSA, 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR