Myosin, the major component of muscle filaments, is a long asymmetric molecule containing a globular head and a long tail. It is composed of two heavy chains and four light chains. Phosphorylation of myosin light chain in response to increased calcium levels leads to contraction. Myosin light chain phosphatase acts to regulate muscle contraction by dephosphorylating activated myosin light chain.
MYL9 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200, immunofluorescence starting at 1:200 - 1:1000, and flow cytometry starting at 1:200 - 1:400.
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: MYL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat