Anti-HDAC4 Rabbit Polyclonal Antibody

Supplier: Prosci

48-701
10078-316EA 582.9 USD
10078-316
Anti-HDAC4 Rabbit Polyclonal Antibody
Antibodies
Histone Deacetylase 4 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200, immunofluorescence starting at 1:200 - 1:1000, and flow cytometry starting at 1:200 - 1:400.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HDAC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR