Cyclin-dependent kinase 2 (CDK2), a CDC2/CDK-type protein kinase, regulates cell-cycle transition from G1 to S phase. CDK2 activation requires formation of a heterodimer with cyclin A, D, or E and phosphorylation by CAK. CDK2 inhibition results from association with p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B). Two alternatively spliced variants and multiple transcription initiation sites have been reported. Inhibition of CDK2 has been suggested to help prevent chemotherapy-induced alopecia in cancer patients.
CDK2 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.
PBS, 0.02% sodium azide, 50% glycerol.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CDK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat