Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARK7 Rabbit Polyclonal Antibody

Supplier: Prosci
48-677
The product of the PARK7 gene, known as PARK7, Parkinson disease (autosomal recessive, early onset) 7, DJ-1 mutant, and DJ-1 oncogene product, functions as a positive regulator of androgen receptor-dependent transcription. PARK7 may also function as a redox-sensitive chaperone and as a sensor for oxidative stress and also has been reported to prevent aggregation of SNCA. PARK7 protects neurons against oxidative stress and cell death and plays a role in fertilization. While PARK7 has no proteolytic activity, it does have a weak transforming activity. PARK7 forms a homodimer and binds to DJBP and PIAS2. PARK7 is part of a ternary complex containing PARK7, DJBP and AR and shows both a nuclear and cytoplasmic localization. In some cells, PARK7 is associated with mitochondria, particularly after oxidative stress. This protein is highly expressed in pancreas, kidney, skeletal muscle, liver, testis and heart tissue, and is expressed at lower levels in placenta and brain. PARK7 is detected in tau inclusions in brains from neurodegenerative disease patients, in astrocytes, Sertoli cells, spermatogonia, spermatids and spermatozoa. Defects in PARK7 are the cause of autosomal recessive early-onset Parkinson's disease 7 (PARK7). Parkinson's disease (PD) is a complex, multifactorial disorder that is characterized by bradykinesia, resting tremor, muscular rigidity and postural instability, response to treatment with levodopa, the loss of dopaminergic neurons in the substantia nigra and the presence of Lewy bodies (intraneuronal accumulations of aggregated proteins) in surviving neurons.

PARK7 antibody can be used in Western Blot, and immunohistochemistry starting at 2.5 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Store PARK7 antibody at -20 °C prior to opening. Dilute only prior to immediate use. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PARK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: African Green Monkey, Chimpanzee, Human, Zebrafish




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 µg 48-677 10078-276 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PARK7
Antigen Name Parkinson Disease Protein 7
Reactivity African Green Monkey, Human, Zebrafish, Chimpanzee
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen Synonyms PARK-7, Parkinsonism associated deglycase, HEL-S-67p, DJ-1, DJ1
Immunogen PARK7 antibody was raised against amino acids 177 - 189 of PARK7 (Human).
Form Liquid
Tested Applications Immunoaffinity Chromatography

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty