Anti-AURKB Rabbit Polyclonal Antibody

48-676
10078-274EA 711.98 USD
10078-274
Anti-AURKB Rabbit Polyclonal Antibody
Antibodies
Aurora Kinase B (also known as Aurora- and Ipl1-like midbody-associated protein 1, AIM-1, Aurora/IPL1-related kinase 2, Aurora-related kinase 2, STK-1, and Aurora-B) is a Ser/Thr protein kinase member of the Aurora subfamily that may be directly involved in regulating the cleavage of polar spindle microtubules and is a key regulator for the onset of cytokinesis during mitosis. Aurora Kinase B is localized to the midzone of central spindle in late anaphase and concentrated into the midbody in telophase and cytokinesis and is colocalized with gamma tubulin in the mid-body. High levels of Aurora B expression are seen in the thymus, although it is also expressed in the spleen, lung, testis, colon, placenta and fetal liver. Aurora B is expressed during S and G2/M phase and expression is up-regulated in cancer cells during M phase.

Aurora Kinase B antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR