Anti-MDH1 Sheep Polyclonal Antibody

Supplier: Prosci
48-671
10076-012EA 785.68 USD
10076-012
Anti-MDH1 Sheep Polyclonal Antibody
Antibodies
MDH1 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR