Anti-DLD Rabbit Polyclonal Antibody

48-668
10076-008EA 674.53 USD
10076-008
Anti-DLD Rabbit Polyclonal Antibody
Antibodies
Dihydrolipoyl dehydrogenase is a component of the mitochondrial pyruvate and alpha-ketoglutarate dehydrogenase complexes. Defects in the enzyme cause congenital infantile lactic acidosis.

DLD antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

IgG Fractionation

PBS, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DLD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR