Anti-CPA1 Rabbit Polyclonal Antibody

48-666
10076-004EA 750.52 USD
10076-004
Anti-CPA1 Rabbit Polyclonal Antibody
Antibodies
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The A1 and A2 forms are monomeric proteins with different biochemical properties. Carboxypeptidase A1 is a monomeric pancreatic exopeptidase. It is involved in zymogen inhibition.

CPA1 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

IgG Fractionation

PBS, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR