Anti-REN Sheep Polyclonal Antibody

48-665
10076-002EA 674.53 USD
10076-002
Anti-REN Sheep Polyclonal Antibody
Antibodies
Renin antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

0.05 M sodium phosphate; 0.1 M NaCl; 1 mM EDTA;, pH 6.6.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: REN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR