Anti-CA2 Rabbit Polyclonal Antibody

Supplier: Prosci

48-664
10076-000EA 582.9 USD
10076-000
Anti-CA2 Rabbit Polyclonal Antibody
Antibodies
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.

Carbonic Anhydrase II antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

IgG Fractionation

PBS, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR