Anti-NCK2 Rabbit Polyclonal Antibody

48-611
10078-248EA 749.47 USD
10078-248
Anti-NCK2 Rabbit Polyclonal Antibody
Antibodies
NCK2 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, and flow cytometry starting at 1:50 - 1:100.

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: NCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR