Anti-SOCS4 Rabbit Polyclonal Antibody

48-585
10078-232EA 749.47 USD
10078-232
Anti-SOCS4 Rabbit Polyclonal Antibody
Antibodies
SOCS4 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200 - 1:1000, and immunofluorescence starting at 1:200 - 1:1000.

PBS, pH 7.4, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: SOCS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR