Anti-NOS3 Rabbit Polyclonal Antibody

48-573
10078-224EA 520.13 USD
10078-224
Anti-NOS3 Rabbit Polyclonal Antibody
Antibodies
NOS3 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200 - 1:1000.

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NOS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR