Anti-MMP23B Rabbit Polyclonal Antibody

Supplier: Prosci

48-566
10078-216EA 624.31 USD InStock
10078-216
Anti-MMP23B Rabbit Polyclonal Antibody
Antibodies
MMP23A and MMP23B, two Metallopeptidase type proteases, are type II membrane-anchored zymogens that are proteolytically processed at a conserved motif and converted into secretory and mature enzymes. The proteins contain prepro, catalytic, cysteine-rich, interleukin-1 receptor-related and proline-rich domains. The MMP23A and MMP23B genes have been reported to express multiple mRNAs, some of which are derived by alternative splicing.

MMP23B antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200, and immunofluorescence starting at 1:200 - 1:1000.

Affinity Chromatography

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: MMP23B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR