Anti-BCL2 Rabbit Polyclonal Antibody

48-506
10078-162EA 540.97 USD InStock
10078-162
Anti-BCL2 Rabbit Polyclonal Antibody
Antibodies
Bcl-2 inhibits apoptosis. Translocations that lead to overexpression of this gene are associated with cancer (lymphoma in the case of translocation to the Ig heavy chain locus).

BCL2 antibody can be used in immunohistochemistry starting at 40 ?g/mL, and flow cytometry.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR