Anti-ADD1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-499
10078-150EA 582.9 USD
10078-150
Anti-ADD1 Rabbit Polyclonal Antibody
Antibodies
ADD1 antibody can be used in Western Blot, immunohistochemistry starting at 1:100, immunoprecipitation, and flow cytometry.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ADD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR