Anti-RREB1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-494
10078-142EA 582.9 USD
10078-142
Anti-RREB1 Rabbit Polyclonal Antibody
Antibodies
RREB1 (also known as RAS-responsive element binding protein 1 and Raf responsive zinc finger protein) is a transcription factor that binds specifically to the distal RAS-responsive element (RRE) in the calcitonin gene promoter and augments the Ras/Raf-mediated transcriptional response of that promoter. RREB1 may be involved in Ras/Raf-mediated cell differentiation. RREB is a nuclear protein expressed in heart, placenta, lung, liver, skeletal muscle, kidney and pancreas and is not found in the brain. The protein contains 4 C2H2-type zinc fingers.

DNA-binding Protein antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

Protein A Column

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR