Anti-CD79a Rabbit Polyclonal Antibody

48-454
10078-122EA 749.47 USD
10078-122
Anti-CD79a Rabbit Polyclonal Antibody
Antibodies
CD79A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% BSA, 0.1% sodium azide.

Aliquot and store at 4°C. Avoid freezing.

Type: Primary
Antigen: CD79A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR