Anti-OAS3 Rabbit Polyclonal Antibody

48-426
10078-100EA 540.97 USD
10078-100
Anti-OAS3 Rabbit Polyclonal Antibody
OAS3 antibody can be used in ELISA starting at 1:10000 - 1:50000, Western Blot starting at 1:1000 - 1:5000, immunohistochemistry starting at 20 ?g/mL, and immunofluorescence starting at 1:200 - 1:1000.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: OAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR