Anti-ITGB1 Rabbit Polyclonal Antibody

48-408
10074-324EA 698.16 USD
10074-324
Anti-ITGB1 Rabbit Polyclonal Antibody
Antibodies
Integrins are a major class of cell adhesion molecules that mediate cell matrix interactions. They are composed of an alpha and a beta subunit. Beta 1 heterodimers are widely expressed and control various developmental processes, including neurogenesis, chondrogenesis, myoblast fusion, and skin and hair follicle morphogenesis.

Integrin beta 1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ITGB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Dog, Gorilla, Horse, Human, Monkey, Orangutan, Pig, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR