Anti-KIT Rabbit Polyclonal Antibody

48-391
10075-970EA 491.23 USD
10075-970
Anti-KIT Rabbit Polyclonal Antibody
KIT encodes the human homolog of the proto-oncogene c-kit. C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). Somatic gain-of-function mutations in the extracellular domain, kinase domains 1 and 2 are found in gastrointestinal stromal tumors (GISTs). Mutations in c-KIT gene are also associated with mast cell disease, acute myelogenous lukemia, and piebaldism. Tyrosine kinase inhibitor STI571 has been shown to inhibit GIST cells that express mutated c-KIT.

KIT Oncogene antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200 - 1:400.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Dog, Gibbon, Goat, Gorilla, Horse, Human, Monkey, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR