Anti-LINGO1 Rabbit Polyclonal Antibody

48-383
10078-068EA 698.16 USD
10078-068
Anti-LINGO1 Rabbit Polyclonal Antibody
Antibodies
Also known as LRRN6A (leucine-rich repeat neuronal 6A), LINGO1 is a newly discovered gene that is expressed primarily in the central nervous system during early embro development, and may be an essential protein in the development of the nervous system.

LINGO1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunofluorescence starting at 1 - 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LINGO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Dog, Gorilla, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR