Anti-USP2 Rabbit Polyclonal Antibody

48-371
10078-046EA 674.53 USD
10078-046
Anti-USP2 Rabbit Polyclonal Antibody
Antibodies
UBP, an ubiquitin-specific protease, is selectively up regulated in bone by the osteotropic agents PTH, PTHrP and PGE2 and possibly via the PKA/cAMP pathway. It is also thought to play a role in the recycling of ubiquitin by hydrolysis of branched poly-ubiquitin from linear poly-ubiquitin chains, production of free ubiquitin from linear poly-ubiquitin chains and from certain ribosomal ubiquitin fusion proteins.

USP2 antibody can be used in ELISA, Western Blot starting at 1:1000 - 1:2000, and immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: USP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR