Anti-ATRN Rabbit Polyclonal Antibody

48-352
10078-008EA 674.53 USD
10078-008
Anti-ATRN Rabbit Polyclonal Antibody
Antibodies
Attractin antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Dog, Hamster, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR