Anti-ATRN Rabbit Polyclonal Antibody

48-351
10078-006EA 674.53 USD
10078-006
Anti-ATRN Rabbit Polyclonal Antibody
Antibodies
Attractin antibody can be used in ELISA starting at 1:10000, and immunohistochemistry starting at 1:200 - 1:400.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR