Anti-DPP4 Rabbit Polyclonal Antibody

48-348
10078-000EA 698.16 USD
10078-000
Anti-DPP4 Rabbit Polyclonal Antibody
Antibodies
DPP4 antibody can be used in immunohistochemistry starting at 1:50, and immunofluorescence.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DPP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR