Anti-PDE3A Rabbit Polyclonal Antibody

48-336
10077-978EA 674.53 USD
10077-978
Anti-PDE3A Rabbit Polyclonal Antibody
Antibodies
Phosphodiesterase 3A is a cGMP inhibited, cAMP/cGMP dual-specific phosphodiesterase. PDE3A has been implicated in male erectile dysfunction, mammalian oocyte maturation, platelet function, vascular smooth muscle function, and salivary gland tumors. Three alternatively spliced transcripts have been reported.

PDE3A antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, immunofluorescence starting at 10 ?g/mL, and immunoprecipitation.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDE3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Horse, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR