Anti-PDE2A Rabbit Polyclonal Antibody

48-327
10077-964EA 674.53 USD
10077-964
Anti-PDE2A Rabbit Polyclonal Antibody
Antibodies
Phosphodiesterase 2A is a cGMP stimulated, cAMP/cGMP dual-specific phosphodiesterase whose hydrolytic activity is stimulated by the presence of cGMP binding to the GAF domain. PDE2A has been implicated in penile erectile dysfunction and in the regulation of fluid and inflammatory cell transit across the endothelial cell barrier (i.e., venous and capillary). PDE2A activity is inhibited by erythro-9-(2-hydroxyl-3-nonyl)adenine (EHNA). At least three isoforms have been reported (PDE2A1, PDE2A2, and PDE2A3).

PDE2A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDE2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gorilla, Horse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR