Anti-DUSP26 Rabbit Polyclonal Antibody

48-304
10077-918EA 674.53 USD
10077-918
Anti-DUSP26 Rabbit Polyclonal Antibody
Antibodies
MGC1136 is classified as a dual specificity MAP kinase phosphatase.

DUSP26 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DUSP26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Orangutan, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR