Anti-GRM2 Rabbit Polyclonal Antibody

48-303
10075-956EA 674.53 USD
10075-956
Anti-GRM2 Rabbit Polyclonal Antibody
Antibodies
The Metabotropic Glutamate Receptor GRM2 binds L-glutamate and has been linked to the inhibition of the cyclic AMP signaling cascade. GRM2 is involved in mediating the suppression of neurotransmission and in synaptogenesis and synaptic stabilization. Glutamatergic neurotransmission plays a role in most aspects of normal brain function and can be perturbed in many neuropathologic conditions.

GRM2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GRM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chimpanzee, Gorilla, Human, Monkey, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR