Anti-GPR61 Rabbit Polyclonal Antibody

48-302
10075-954EA 698.16 USD
10075-954
Anti-GPR61 Rabbit Polyclonal Antibody
Antibodies
GPR61 is an Orphan-A GPCR with an unknown ligand.

GPR61 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Gorilla, Horse, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR