Anti-FFAR2 Rabbit Polyclonal Antibody

48-292
10075-934EA 698.16 USD
10075-934
Anti-FFAR2 Rabbit Polyclonal Antibody
Antibodies
GPR43 is classified as a Carboxylic Acid GPCR. The location of this receptor in the immune system is suggestive of potential roles in immune cell function and hematopoiesis.

FFAR2 antibody can be used in immunohistochemistry starting at 7 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FFAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR