Anti-FZD7 Rabbit Polyclonal Antibody

Supplier: Prosci
48-270
10075-890EA 785.68 USD
10075-890
Anti-FZD7 Rabbit Polyclonal Antibody
Antibodies
FZD7, a Frizzled Receptor, is a receptor for Wnt signaling proteins. FZD7 downregulates the functions of the tumor suppressor gene APC and thereby enhances beta catenin-mediated signals.

Frizzled 7 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FZD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Dog, Gorilla, Hamster, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR