Anti-CCR4 Rabbit Polyclonal Antibody

48-261
10075-872EA 684.46 USD
10075-872
Anti-CCR4 Rabbit Polyclonal Antibody
Antibodies
CCR4, a Chemokine Receptor, mediates inflammatory responses and HIV-1 infections. It binds beta-family chemokines (CC motif) such as TARC (thymus and activation-regulated chemokine) and MDC (macrophage-derived chemokine).

CCR4 antibody can be used in immunohistochemistry starting at 6 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR