Anti-GPR77 Rabbit Polyclonal Antibody

48-256
10075-862EA 698.16 USD
10075-862
Anti-GPR77 Rabbit Polyclonal Antibody
Antibodies
C5L2 is an Anaphylatoxin Receptor. Binding to anaphylatoxins can result in increased triglyceride synthesis, although its precise role in cellular responses has not been determined.

GPR77 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR