Anti-LPAR5 Rabbit Polyclonal Antibody

48-253
10075-856EA 684.46 USD
10075-856
Anti-LPAR5 Rabbit Polyclonal Antibody
Antibodies
GPR92/GPR93 is an Orphan-A GPCR with an unknown ligand.

GPR92 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LPAR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR