Prostaglandin E Receptor EP2 is a member of the Prostanoid Receptor subfamily. Along with several other receptor subtypes (EP1, EP3, and EP4), it mediates the diverse biological effects of Prostaglandin E2 (PGE2). PGE2 has been implicated in numerous processes, including immune response modulation, male erectile function, induction of labor, cervical cancer, control of blood pressure, asthma, and more recently, Alzheimer's Disease.
PTGER2 antibody can be used in ELISA, and immunohistochemistry starting at 20 ?g/mL.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PTGER2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Gorilla, Human, Mouse, Pig