Anti-GPRC5B Rabbit Polyclonal Antibody

48-246
10075-842EA 684.46 USD
10075-842
Anti-GPRC5B Rabbit Polyclonal Antibody
Antibodies
GPRC5B is an Orphan-C GPCR induced by retinoids. GPRC5B shows some homologies to the metabotropic glutamate receptor-like family of GPCRs; however, the GPRC5B protein has a short N-terminal domain compared to other family C members.

GPRC5B antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPRC5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Horse, Human, Monkey, Orangutan, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR